Learn More
Sino Biological Anti-EGFR Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10001MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EGFR / HER1 / ErbB1 (rh EGFR / HER1 / ErbB1; Catalog#10001-H20B2; NP_005219; Met668-Ala1210). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0258