Learn More
Sino Biological Recombinant Anti-EGFR Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10001RE01A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EGFR / HER1 / ErbB1 (rh EGFR / HER1 / ErbB1; Catalog#10001-H08H; NP_005219; Met1-Ser645) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-0269