Learn More
Sino Biological EGFR / HER1 / ErbB1 Neutralizing Antibody
Supplier:  Sino Biological 10001REN01200
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EGFR / HER1 / ErbB1 (rh EGFR / HER1 / ErbB1; Catalog#10001-H08H; NP_005219; Met1-Ser645) and was produced using recombinant antibody technology.
Catalog No. 50-161-0279