Learn More
Sino Biological Influenza A H5N1 (A/VietNam/1203/2004) Hemagglutinin Protein (HA1 Subunit) (His & Fc Tag)
Supplier:  Sino Biological 10003V06H1100
Encompass_Preferred
A DNA sequence encoding an N-terminal segment (Met 1-Arg 342) of Influenza A Virus H5N1 Hemagglutinin (AAW80717.1),termed as HA1, was fused with the C-terminal Fc region of mouse IgG1 followed by a polyhistidine tag.
Catalog No. 50-161-3733