Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10004H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 652) of human ErbB2 (NP_004439.2) was fused with the polyhistidine tag at the C-terminus.
Catalog No. 50-161-3740