Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (ECD), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10004HCCHB20
Encompass_Preferred
A DNA sequence encoding the human ERBB2 (NP_004439.2) (Met1-Thr652) was expressed with five amino acids (DDDDK) at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-161-3753