Learn More
Sino Biological Anti-Her2/ERBB2 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10004MM03F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ErbB2 / HER2 / CD340 (rh ErbB2 / HER2 / CD340; Catalog#10004-H08H; NP_004439.2; Met 1-Thr 652) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-0333