Learn More
Sino Biological Recombinant Anti-Her2/ERBB2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10004R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ErbB2 / HER2 cytoplasmic domain (rhErbB2; Catalog#10004-H20B1; NP_004439.2; Lys 676-Val 1255).
Catalog No. 50-161-0346