Learn More
Sino Biological Recombinant Anti-Her2/ERBB2 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10004R511A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ErbB2 / HER2 extracellular domain (rh ErbB2; Catalog#10004-H08H; NP_004439.2; Met 1-Thr 652) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-0355