Learn More
Sino Biological Recombinant Human Nectin-2 Protein (hFc Tag)
Supplier:  Sino Biological 10005H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Leu 360) of human CD112 (NP_002847.1) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-3754