Learn More
Sino Biological Recombinant Human G-CSF Protein (Isoform b), HPLC-verified
Supplier:  Sino Biological 10007HNAH100
Encompass_Preferred
A DNA sequence encoding the human CSF3 (NP_757373.1) (Met1-Pro204) was expressed with an initial Met.
Catalog No. 50-161-3763