Learn More
Sino Biological VEGF121 Protein, Human, Recombinant
Supplier:  Sino Biological 10008HNAH10
Encompass_Preferred
A DNA sequence encoding the human VEGF 121 isoform (P15692-9) (Met1-Arg147) was expressed.
Catalog No. 50-161-3776