Learn More
Sino Biological VEGF183 Protein, Human, Recombinant
Supplier:  Sino Biological 10009HNAB50
Encompass_Preferred
A DNA sequence encoding the human VEGF 183 isoform (P15692-3) (Met1-Arg209) was expressed.
Catalog No. 50-161-3779