Learn More
Sino Biological Recombinant Anti-IL-1RA/IL-1RN Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100099R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the human IL1Ra / IL1RN.
Catalog No. 50-161-0436