Learn More
C&G Containers 1000 mL Wide-Mouth Oblong Bottle, Excellent Moisture Barrier, High Impact Resistance, 1000 mL
SDP
Catalog No. NC2191297
Encompass_Preferred
The 1000 mL wide-mouth oblong bottle is crafted from natural HDPE, featuring a durable design ideal for scientific and industrial use. Its excellent moisture barrier and high impact resistance ensure safe storage of solids and powders, while the 53-400 neck finish allows for easy filling and dispensing. Perfect for pharmaceuticals and transport, this bottle's space-saving shape makes it an efficient choice for laboratory settings.
This item is not returnable. View return policy
Catalog No. NC2191297 Supplier C&G Containers Supplier No. LPO032500013
The 1000 mL wide-mouth oblong bottle is crafted from natural HDPE, featuring a durable design ideal for scientific and industrial use. Its excellent moisture barrier and high impact resistance ensure safe storage of solids and powders, while the 53-400 neck finish allows for easy filling and dispensing. Perfect for pharmaceuticals and transport, this bottle's space-saving shape makes it an efficient choice for laboratory settings.