Learn More
S2 Media 1L 10x TRIS Buffered Saline (TBS, 10x), pH 7.4 in 1L Media Bottle, 1/each
Catalog No. 502995857
Encompass_Preferred
S2Media™ 10x TRIS Buffered Saline (TBS, 10x), pH 7.4 is a ready to use stock buffer often used for immunodetection techniques, including Western blot protocols. To prepare a 1x solution, mix one volume of 10x solution with 9 volumes of 10x buffer solution.• Product meets reference methods and performance criteria.• TBS,10x should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions. • Product is 0.1µm filtered. Bottles are RNase, DNase, and protease free.
Catalog No. 50-299-5857 Supplier S2 Media Supplier No. 5354
Add to Cart
Add to Cart
S2Media™ 10x TRIS Buffered Saline (TBS, 10x), pH 7.4 is a ready to use stock buffer often used for immunodetection techniques, including Western blot protocols. To prepare a 1x solution, mix one volume of 10x solution with 9 volumes of 10x buffer solution.• Product meets reference methods and performance criteria.• TBS,10x should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions. • Product is 0.1µm filtered. Bottles are RNase, DNase, and protease free.