Learn More
S2 Media 1M Tris-HCl, pH 8.0, Sterile, 1L Media Bottle, 1ea
Catalog No. 502995853
Encompass_Preferred
1M Tris-HCl Buffer (sterile) is commonly used in labs for pH stabilization in biochemical reactions, as a buffer in analytical chemistry, and as a sample diluent near neutral to slightly basic pH. At pH 8.0, Tris (pKa ≈ 8.1 at 25°C) provides effective buffering between pH 7–9, making it compatible with most biological systems and minimally reactive with biomolecules.• 1M Tris-HCl should be used by trained professionals within a laboratory setting. • Tris is an abbreviation for tris(hydroxymethyl)aminomethane).• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• For Research Use Only (RUO). Not for use in diagnostic procedures.
Catalog No. 50-299-5853 Supplier S2 Media Supplier No. 5359
1M Tris-HCl Buffer (sterile) is commonly used in labs for pH stabilization in biochemical reactions, as a buffer in analytical chemistry, and as a sample diluent near neutral to slightly basic pH. At pH 8.0, Tris (pKa ≈ 8.1 at 25°C) provides effective buffering between pH 7–9, making it compatible with most biological systems and minimally reactive with biomolecules.• 1M Tris-HCl should be used by trained professionals within a laboratory setting. • Tris is an abbreviation for tris(hydroxymethyl)aminomethane).• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• For Research Use Only (RUO). Not for use in diagnostic procedures.