Learn More
S2 Media 1L 1x Phosphate Buffered Saline (PBS), pH 6.0, 1L Media Bottle, 1ea
Catalog No. 502995734
Encompass_Preferred
S2Media™ 1x Phosphate Buffered Saline (PBS), pH 6.0 has many applications due to its isotonic nature, including uses as a rinse fluid, diluent, and buffer. Phosphate salts are nontoxic to living cells and have a high buffering capacity. pH maintenance is important in retaining cell viability and in the recovery and revitalization of injured or damaged cells during bacterial culture and analysis. PBS is generally used as a 10mM working solution containing a mixture of sodium chloride and inorganic phosphate salts.• Product meets AOAC, APHA, BAM, CLSI, SM-Dairy, SMWW, and USP-NF reference methods and performance criteria.• For in vitro diagnostic (IVD) use.• PBS should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5734 Supplier S2 Media Supplier No. 5355
S2Media™ 1x Phosphate Buffered Saline (PBS), pH 6.0 has many applications due to its isotonic nature, including uses as a rinse fluid, diluent, and buffer. Phosphate salts are nontoxic to living cells and have a high buffering capacity. pH maintenance is important in retaining cell viability and in the recovery and revitalization of injured or damaged cells during bacterial culture and analysis. PBS is generally used as a 10mM working solution containing a mixture of sodium chloride and inorganic phosphate salts.• Product meets AOAC, APHA, BAM, CLSI, SM-Dairy, SMWW, and USP-NF reference methods and performance criteria.• For in vitro diagnostic (IVD) use.• PBS should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.