Learn More
S2 Media 20x Tris Buffered Saline (400mM Tris-HCl, 3M NaCl), pH 7.4, Sterile, 1L Media Bottle, 1ea
Catalog No. 502995854
Encompass_Preferred
Tris Buffered Saline (TBS), formulated at a 20x concentration, is used in cell culture, molecular biology, protein research, and purification applications. It can be utilized as a wash buffer, storage buffer, and for resuspension of cells. he Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical applications. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration.• Tris Buffered Saline should be used by trained professionals within a laboratory setting. • Tris is an abbreviation for tris(hydroxymethyl)aminomethane).• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• For Research Use Only (RUO). Not for use in diagnostic procedures.
Catalog No. 50-299-5854 Supplier S2 Media Supplier No. 5360
Tris Buffered Saline (TBS), formulated at a 20x concentration, is used in cell culture, molecular biology, protein research, and purification applications. It can be utilized as a wash buffer, storage buffer, and for resuspension of cells. he Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical applications. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration.• Tris Buffered Saline should be used by trained professionals within a laboratory setting. • Tris is an abbreviation for tris(hydroxymethyl)aminomethane).• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• For Research Use Only (RUO). Not for use in diagnostic procedures.