Learn More
Ace Glass, Inc. 1000mL 24/25 separatory funnel with polyethylene stopper
SDP
Catalog No. 50974692
Encompass_Preferred
1000ml separatory funnel with polyethylene stopper. Outer ground joint at top is a 24/25 Rodaviss joint. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-692 Supplier Ace Glass, Inc. Supplier No. 723118
1000ml separatory funnel with polyethylene stopper. Outer ground joint at top is a 24/25 Rodaviss joint. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.