Learn More
Ace Glass, Inc. 1000mL graduated cylindrical funnel, 24/40 top, 24/40 bottom
SDP
Catalog No. 50979234
Encompass_Preferred
1000mL graduated cylindrical funnel with 24/40 joints at top and bottom. Features include drip stem, and double scale graduations calibrated from stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Includes PTFE stopcock plug.
Catalog No. 50-979-234 Supplier Ace Glass, Inc. Supplier No. 726866
1000mL graduated cylindrical funnel with 24/40 joints at top and bottom. Features include drip stem, and double scale graduations calibrated from stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Includes PTFE stopcock plug.