Learn More
Chemglass Life Sciences 1000mL Flask, Reaction, 24/40 Outer Joint, 0-8mm Valve, Airfree, Schlenk
SDP
Catalog No. AF052907
Encompass_Preferred
1000mL Flask, Reaction, 24/40 Outer Joint, 0-8mm Valve, Airfree, Schlenk. Round bottom single neck flask having a standard taper outer joint with a Chem-Vac™ Chem-Cap valve sidearm. Plug may be removed for sampling from flask while a purge of inert gas is maintained thru the sidearm. Valve size is as listed.
Catalog No. AF052907 Supplier Chemglass Life Sciences Supplier No. AF052907
1000mL Flask, Reaction, 24/40 Outer Joint, 0-8mm Valve, Airfree, Schlenk. Round bottom single neck flask having a standard taper outer joint with a Chem-Vac™ Chem-Cap valve sidearm. Plug may be removed for sampling from flask while a purge of inert gas is maintained thru the sidearm. Valve size is as listed.