Learn More
Ace Glass, Inc. 1000mL 24/40 separatory funnel body only
SDP
Catalog No. 50979220
Encompass_Preferred
1000ml separatory funnel with 24/40 outer ground Rodaviss joint at top. Funnel body supplied with stopcock plug.
Catalog No. 50-979-220 Supplier Ace Glass, Inc. Supplier No. 723142
1000ml separatory funnel with 24/40 outer ground Rodaviss joint at top. Funnel body supplied with stopcock plug.