Learn More
Chemglass Life Sciences 1000mL 3-Neck European Flask, Jacketed, 1-CN 24/40 Outer, 2-SN 24/40 Outer Angled 20°
SDP
Catalog No. 50872080
Encompass_Preferred
1000mL 3-Neck European Flask, Jacketed, 1-CN 24/40 Outer, 2-SN 24/40 Outer Angled 20¿. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. Entire body of the flask is jacketed and has two hose connections for circulation of heating or cooling liquid. With standard taper outer joints on center and side necks.
Catalog No. 50-872-080 Supplier Chemglass Life Sciences Supplier No. CG157608
1000mL 3-Neck European Flask, Jacketed, 1-CN 24/40 Outer, 2-SN 24/40 Outer Angled 20¿. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. Entire body of the flask is jacketed and has two hose connections for circulation of heating or cooling liquid. With standard taper outer joints on center and side necks.