Learn More
Chemglass Life Sciences 1000mL Addition Funnel, 24/40 Joint Size, 0-4mm PTFE Valve, 545mm OAH
SDP
Catalog No. CG170406
Encompass_Preferred
1000mL Addition Funnel, 24/40 Joint Size, 0-4mm PTFE Valve, 545mm OAH. Cylindrical style addition funnel with a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. For replacement stoppers see CG-3000.
Catalog No. CG170406 Supplier Chemglass Life Sciences Supplier No. CG170406
1000mL Addition Funnel, 24/40 Joint Size, 0-4mm PTFE Valve, 545mm OAH. Cylindrical style addition funnel with a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. For replacement stoppers see CG-3000.