Learn More
Bellco Glass, Inc. 1000mL Bottle Top Filter
SDP
Supplier:  Bellco Glass, Inc. 229714000
Encompass_Preferred
Bottle top filters do not include solution bottlestoread molded graduations marks on bottle top filter. Bottle top filters fit on CELLTREAT solution bottles or other standard 45mm threaded solution storage bottles. Polyethersulfone PES membrane is for high flow and low binding. Nylon PA membrane has excellent chemical compatibility and does not require prewetting. Angled hose barb adapter fits three size hose diameters and prevents the filter from tipping.Specifications Easytoread molded graduations marks on bottle top filter Bold easytoread printed membrane and pore size for easy identification Printed lot number and expiration date on every bottle top Improved filter support provides faster flow rates Angled hose barb adapter fits three size hose diameters and prevents the filter from tipping 0.22µm PES membrane is the first choice for sterile filtration of cell culture
Catalog No. 50-232-9740