Learn More
CELLTREAT Scientific Products 1000mL Bottle Top Filter, 0.45m PES Filter, 90mm Diameter, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229714
Encompass_Preferred
CELLTREAT Scientific Products 1000mL, 0.45¿m PES Bottle Top Filter. Easy-to-read molded graduations marks on bottle top filter. Bold easy-to-read printed membrane and pore size for easy identification. Curved bottle design for easy handling. Printed lot number and expiration date on every bottle top. Improved filter support provides faster flow rates. Angled hose barb adapter fits three size hose diameters and prevents the filter from tipping. Polyethersulfone (PES) membrane is for high flow and low binding. 0.45¿m PES membrane ideal for solution clarification. Bottle top filters fit on CELLTREAT solution bottles or other standard 45mm threaded solution storage bottles. Membrane diameter 90mm. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 24/case.
Catalog No. 50-202-050