Learn More
Chemglass Life Sciences Cylinder, 1000mL, Class A, Certified, Single Scale
SDP
Catalog No. NC0976866
Encompass_Preferred
Cylinder, 1000mL, Class A, Certified, Single Scale, Calibrated ¿To Deliver¿ in accordance with ASTM E-1272 and ASTM E-542. Each cylinder is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. Permanent white graduations are easy to read. Bumper guards are supplied with 25mL through 2L inclusive.
Catalog No. NC0976866 Supplier Chemglass Life Sciences Supplier No. CG82561L
Cylinder, 1000mL, Class A, Certified, Single Scale, Calibrated ¿To Deliver¿ in accordance with ASTM E-1272 and ASTM E-542. Each cylinder is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. Permanent white graduations are easy to read. Bumper guards are supplied with 25mL through 2L inclusive.