Learn More
S2 Media 1L D/E (Dey-Engley) Neutralizing Broth without Tween in 1L Media Bottle, 1ea
Catalog No. 502995751
Encompass_Preferred
D/E (Dey-Engley) Neutralizing Broth without Tween 80 is utilized for the neutralization and testing of antiseptics and disinfectants according to the procedures of Engley and Dey. The broth contains a high concentration of lecithin (7.0g/L) and is opaque in appearance. Therefore, turbidity alone cannot be used as a visual indication of growth and subculture onto a suitable solid media is recommended. Organisms that ferment dextrose will cause a color shift from purple to yellow. Species that do not ferment dextrose can form pellicles on the surface of the medium. • Product meets AAMI, APHA, ISO, SM-Dairy, and USP-NF reference methods and performance criteria.• D/E Neutralizing Broth should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Product does not contain polysorbate 80.
This item is not returnable. View return policy
Catalog No. 50-299-5751 Supplier S2 Media Supplier No. 5255
D/E (Dey-Engley) Neutralizing Broth without Tween 80 is utilized for the neutralization and testing of antiseptics and disinfectants according to the procedures of Engley and Dey. The broth contains a high concentration of lecithin (7.0g/L) and is opaque in appearance. Therefore, turbidity alone cannot be used as a visual indication of growth and subculture onto a suitable solid media is recommended. Organisms that ferment dextrose will cause a color shift from purple to yellow. Species that do not ferment dextrose can form pellicles on the surface of the medium. • Product meets AAMI, APHA, ISO, SM-Dairy, and USP-NF reference methods and performance criteria.• D/E Neutralizing Broth should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Product does not contain polysorbate 80.