Learn More
Cole-Parmer 1000 mL Dipper,Polypropylene, with Extendable Handle up to 9 ft
Catalog No. NC1257343 Shop All Cole-Parmer Products
Encompass
Scienceware F37807-0000 1000 mL Dipper, Polypropylene, with Extendable Handle up to 9 ft. For improved sampling at an elevated position, a 1000-mL graduated polypropylene (PP) pitcher is fixed with a downward offset at the end of an aluminum handle. The telescoping handle expands from a minimum length of 3 ft (91 cm) to a maximum length of 9 ft (2.7 m) at 6 in (15 cm) intervals for optimal sampling distance versatility. The three sections of the handle can be disassembled for complete cleaning and easy storage.
Catalog No. NC1257343 Supplier Cole-Parmer Supplier No. 0628290
 Restricted Item
Add to Cart
Add to Cart
Scienceware F37807-0000 1000 mL Dipper, Polypropylene, with Extendable Handle up to 9 ft. For improved sampling at an elevated position, a 1000-mL graduated polypropylene (PP) pitcher is fixed with a downward offset at the end of an aluminum handle. The telescoping handle expands from a minimum length of 3 ft (91 cm) to a maximum length of 9 ft (2.7 m) at 6 in (15 cm) intervals for optimal sampling distance versatility. The three sections of the handle can be disassembled for complete cleaning and easy storage.