Learn More
Chemglass Life Sciences 1000mL Erlenmeyer Flask, #32 Outer Stopper Neck, Graduated, Complete
SDP
Catalog No. CG154613
Encompass_Preferred
1000mL Erlenmeyer Flask, #32 Outer Stopper Neck, Graduated, Complete. Erlenmeyer flask with standard taper stopper top. Available with or without stopper. For stoppers only see CG-3018.
Catalog No. CG154613 Supplier Chemglass Life Sciences Supplier No. CG154613
1000mL Erlenmeyer Flask, #32 Outer Stopper Neck, Graduated, Complete. Erlenmeyer flask with standard taper stopper top. Available with or without stopper. For stoppers only see CG-3018.