Learn More
CELLTREAT Scientific Products 1000mL Erlenmeyer Flask, Vent Cap, Baffled Bottom, PC, Sterile
SDP
Catalog No. NC2206343
Encompass_Preferred
Lightweight plastic material is easier and safer to handle than glass. Designed for mammalian suspension culture applications as well as bacterial & yeast culture. Baffled flasks enhance both vertical and circular culture movement to improve aeration. Vent cap options are available to prevent the common mistake of unintentionally sealing the cap completely and preventing gas exchange. All flasks include molded graduation lines and feature a molded 1/3 fill line to help ensure optimal working volumes. Individually bagged for easy storage and handling. Medical grade virgin Polycarbonate is autoclavable (temperature range of -80°C – 121°C). Working volumes are recommendations only; optimize as required for individual protocols.
This item is not returnable. View return policy
Catalog No. NC2206343 Supplier CELLTREAT Scientific Products Supplier No. 229835
Lightweight plastic material is easier and safer to handle than glass. Designed for mammalian suspension culture applications as well as bacterial & yeast culture. Baffled flasks enhance both vertical and circular culture movement to improve aeration. Vent cap options are available to prevent the common mistake of unintentionally sealing the cap completely and preventing gas exchange. All flasks include molded graduation lines and feature a molded 1/3 fill line to help ensure optimal working volumes. Individually bagged for easy storage and handling. Medical grade virgin Polycarbonate is autoclavable (temperature range of -80°C – 121°C). Working volumes are recommendations only; optimize as required for individual protocols.