Learn More
Bellco Glass, Inc. 1000mL Erlenmeyer Flask Solid
SDP
Catalog No. 502329964
Encompass_Preferred
Designed for mammalian suspension culture applications as well as bacterial amp yeast culture. Vent caps feature 0.22µm hydrophobic membrane which allows for gas exchange without contamination oversized vent area in cap improves gas exchange.Specifications Packaged sterile in individual bags for easy storage and handling Manufactured from medical grade virgin PETG which reduces gas permeability Size 250mL Cap Type Solid Bottom Type Plain Max Volume 265mL Case Quantity 12
Catalog No. 50-232-9964 Supplier Bellco Glass, Inc. Supplier No. 229812000
Designed for mammalian suspension culture applications as well as bacterial amp yeast culture. Vent caps feature 0.22µm hydrophobic membrane which allows for gas exchange without contamination oversized vent area in cap improves gas exchange.Specifications Packaged sterile in individual bags for easy storage and handling Manufactured from medical grade virgin PETG which reduces gas permeability Size 250mL Cap Type Solid Bottom Type Plain Max Volume 265mL Case Quantity 12