Learn More
Sigma Aldrich Fine Chemicals Biosciences EX-CELL(R) 293 Serum-Free Medium for HEK 293 Cells Serum-free, Animal-protein free, without L-glutamine, liquid, suitable for cell culture | MFCD01634638 | 1000ML
Catalog No. NC9469221
Encompass
EX-CELL(R) 293 Serum-Free Medium for HEK 293 Cells Serum-free, Animal-protein free, without L-glutamine, liquid, suitable for cell culture | MFCD01634638 | 1000ML
Catalog No. NC9469221 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 14571C1000ML
EX-CELL(R) 293 Serum-Free Medium for HEK 293 Cells Serum-free, Animal-protein free, without L-glutamine, liquid, suitable for cell culture | MFCD01634638 | 1000ML