Learn More
Chemglass Life Sciences Glas-col Mantle Extension Support for Series inMin, Mantle Size 1000mL, 194mm ID
SDP
Catalog No. CG1100403
Encompass_Preferred
Glas-col Mantle Extension Support for Series inMin, Mantle Size 1000mL, 194mm ID. Glas-Col extension support for use with a smaller Series M mantle. Attached to a stable ring stand or distillation rack, the support can be adjusted to any convenient height. Support is painted gray and has a 6in long, 1/2in O.D. rod.
Catalog No. CG1100403 Supplier Chemglass Life Sciences Supplier No. CG1100403
Glas-col Mantle Extension Support for Series inMin, Mantle Size 1000mL, 194mm ID. Glas-Col extension support for use with a smaller Series M mantle. Attached to a stable ring stand or distillation rack, the support can be adjusted to any convenient height. Support is painted gray and has a 6in long, 1/2in O.D. rod.