Learn More
Chemglass Life Sciences 1000mL Glas-Col Heating Mantle, Series inOin, Cylindrical, 4 3/16in ID x 6 7/8in Depth, Bottom Drain Opening, 400 Watts
SDP
Catalog No. CG1000912
Encompass_Preferred
1000mL Glas-col Heating Mantle, Series O, Cylindrical, 4 3/16in ID x 6 7/8in Depth, Bottom Drain Opening, 400 Watts. This Glas-Col Series O mantle is similar to CG-10007 except mantle is slit lengthwise to a 1 hole in the bottom center with lacing closure to accommodate a cylindrical reaction vessel, CG-1924, with a bottom drain. Mantle is glass fabric, 115 volts, and is supplied with a separable 4 foot, 2 wire extension cord with locking connector for each circuit. The 10,000mL size has two circuits at the listed wattage.
Catalog No. CG1000912 Supplier Chemglass Life Sciences Supplier No. CG1000912
1000mL Glas-col Heating Mantle, Series O, Cylindrical, 4 3/16in ID x 6 7/8in Depth, Bottom Drain Opening, 400 Watts. This Glas-Col Series O mantle is similar to CG-10007 except mantle is slit lengthwise to a 1 hole in the bottom center with lacing closure to accommodate a cylindrical reaction vessel, CG-1924, with a bottom drain. Mantle is glass fabric, 115 volts, and is supplied with a separable 4 foot, 2 wire extension cord with locking connector for each circuit. The 10,000mL size has two circuits at the listed wattage.