Learn More
Ace Glass, Inc. 1000mL filter flask, #25 Ace-Thred top, Ace-Safe hose connection
SDP
Catalog No. 50971344
Encompass_Preferred
1000mL filter flask featuring a #25 Ace-Thred top and an Ace-Safe hose connection. For use with ACE 3702 filtration apparatus.
Catalog No. 50-971-344 Supplier Ace Glass, Inc. Supplier No. 370208
1000mL filter flask featuring a #25 Ace-Thred top and an Ace-Safe hose connection. For use with ACE 3702 filtration apparatus.