Learn More
Chemglass Life Sciences Dewar Flask, Wide Mouth, Metal Housing, Carrying Handle, 1000mL Capacity, 70mm ID X 302mm Inside Depth, 86mm OD X 337mm Height
SDP
Catalog No. CG159501
Encompass_Preferred
Dewar Flask, Wide Mouth, Metal Housing, Carrying Handle, 1000mL Capacity, 70mm ID X 302mm Inside Depth, 86mm OD X 337mm Height. Similar to CG-1594, but with the addition of a convenient carrying handle. Supplied complete with a vented polyethylene stopper to minimize evaporation. Exterior dimensions listed do not include carrying handle.
Catalog No. CG159501 Supplier Chemglass Life Sciences Supplier No. CG159501
Dewar Flask, Wide Mouth, Metal Housing, Carrying Handle, 1000mL Capacity, 70mm ID X 302mm Inside Depth, 86mm OD X 337mm Height. Similar to CG-1594, but with the addition of a convenient carrying handle. Supplied complete with a vented polyethylene stopper to minimize evaporation. Exterior dimensions listed do not include carrying handle.