Learn More
Chemglass Life Sciences Flask, Shake, 1000mL, 3 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 130 x 215
SDP
Catalog No. 501215182
Encompass_Preferred
Flask, Shake, 1000mL, 3 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 130 x 215. Flask has 3 baffles on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker. The polypropylene vented caps have a 0.22µm (PTFE) hydrophobic membrane to allow sterile gas exchange while preventing contamination. Since these caps are vented, the flasks can be autoclaved with the cap attached.
Catalog No. 50-121-5182 Supplier Chemglass Life Sciences Supplier No. CLS205108
Flask, Shake, 1000mL, 3 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 130 x 215. Flask has 3 baffles on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker. The polypropylene vented caps have a 0.22µm (PTFE) hydrophobic membrane to allow sterile gas exchange while preventing contamination. Since these caps are vented, the flasks can be autoclaved with the cap attached.