Learn More
Hach Company 1000mL Flask-Volumet-Glass
Supplier:  Hach Company 1457453
Encompass
Flask-Volumetric-Class A-Glass-1000 mLCalibrated To Contain and manufactured to ASTM E438 Glass in Laboratory Apparatus requirements
Catalog No. 50-002-57129