Learn More
S2 Media 1L Fluid K, 1L Media Bottle, 1/each
Catalog No. 502995741
Encompass_Preferred
Fluid K is utilized as a rinse solution or diluent for sterility testing on samples that contain petrolatum or oil. It can also be used in bioburden testing to rinse membranes and dilute/dissolve samples. Fluid K can also be utilized for rinsing pathways of medical devices and for samples that are difficult to dissolve. • Product meets Harmonized EP/JP/USP reference methods and performance criteria.• Screw cap with septum available. (Email request to info@s2cm.com.)• Fluid K should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5741 Supplier S2 Media Supplier No. 5166
Fluid K is utilized as a rinse solution or diluent for sterility testing on samples that contain petrolatum or oil. It can also be used in bioburden testing to rinse membranes and dilute/dissolve samples. Fluid K can also be utilized for rinsing pathways of medical devices and for samples that are difficult to dissolve. • Product meets Harmonized EP/JP/USP reference methods and performance criteria.• Screw cap with septum available. (Email request to info@s2cm.com.)• Fluid K should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.