Learn More
Chemglass Life Sciences Bottle, 1000mL, Gas Washing, Complete, 34/28, Extra Coarse Fritted Disc
SDP
Catalog No. CG112001
Encompass_Preferred
Bottle, 1000mL, Gas Washing, Complete, 34/28, Extra Coarse Fritted Disc. Large capacity, low form gas washing bottle having a 34/28 standard taper top outer joint. Filter candle is 25mm O.D. x 70mm long allowing a large gas flow to be dispersed throughout the absorbing material. EXTRA COARSE porosity filter candle. Complete bottle is supplied with one each CG-145-07 size 34 Keck clamp.
Catalog No. CG112001 Supplier Chemglass Life Sciences Supplier No. CG112001
Bottle, 1000mL, Gas Washing, Complete, 34/28, Extra Coarse Fritted Disc. Large capacity, low form gas washing bottle having a 34/28 standard taper top outer joint. Filter candle is 25mm O.D. x 70mm long allowing a large gas flow to be dispersed throughout the absorbing material. EXTRA COARSE porosity filter candle. Complete bottle is supplied with one each CG-145-07 size 34 Keck clamp.