Learn More
Chemglass Life Sciences Cylinder, Large Capacity, 1000mL, Polypropylene, Graduated
SDP
Catalog No. CG122420
Encompass_Preferred
Cylinder, Large Capacity, 1000mL, Polypropylene, Graduated. The handle on the Holdfast™ graduated cylinder makes pouring safer and more convenient. The handle is generously sized for large hands but with a slender enough grip for smaller hands. Hand opening is 3 1/2” x 1 1/8”. Cylinders have a double scale and hex base for stability. Steam autoclavable at 121º C.
Catalog No. CG122420 Supplier Chemglass Life Sciences Supplier No. CG122420
Cylinder, Large Capacity, 1000mL, Polypropylene, Graduated. The handle on the Holdfast™ graduated cylinder makes pouring safer and more convenient. The handle is generously sized for large hands but with a slender enough grip for smaller hands. Hand opening is 3 1/2” x 1 1/8”. Cylinders have a double scale and hex base for stability. Steam autoclavable at 121º C.