Learn More
Chemglass Life Sciences 1000mL Single Neck Round Bottom Flask (RBF) , 29/42 Outer Joint
SDP
Catalog No. CG150624
Encompass_Preferred
1000mL Single Neck Round Bottomed Flask, 29/42 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. CG150624 Supplier Chemglass Life Sciences Supplier No. CG150624
1000mL Single Neck Round Bottomed Flask, 29/42 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.