Learn More
NEST SCIENTIFIC INC 1000ml PETG Erlenmeyer Flask, Flat base, Vent Cap, Sterile,6/cs
Catalog No. NC2887689
Encompass_Preferred
Nest Scientific PETG shaker flasks are ideal for growing suspended cell lines. Molded from USP grade VI virgin PETG. Flasks feature a non-wettable 0.22um PTFE membrane for sterile gas exchange. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.
Catalog No. NC2887689 Supplier NEST SCIENTIFIC INC Supplier No. 784011
Nest Scientific PETG shaker flasks are ideal for growing suspended cell lines. Molded from USP grade VI virgin PETG. Flasks feature a non-wettable 0.22um PTFE membrane for sterile gas exchange. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.