Learn More
Bellco Glass, Inc. 1000mL Polypropylene BottleTop
SDP
Supplier:  Bellco Glass, Inc. 229736000
Encompass_Preferred
CELLTREAT Polypropylene Bottle FiltersSpecifications Fits on standard GL 45 threaded glass vacuum bottles Excellent chemical resistance offered with polypropylene housing as compared to polystyrene High temperature range of 4˚C to 37˚C is suitable for many applications Hydrophilic nylon membrane has excellent chemical compatibility with esters bases and alcohols and does not require prewetting Hydrophobic PTFE membrane has excellent temperature resistance and is useful for clarifying aqueous samples as well as strong acid and alkali solvent filtration Membrane Diameter 90 Filter Pore Size 0.45um Case Quantity 12
Catalog No. 50-233-0095