Learn More
CELLTREAT Scientific Products 1000mL Polypropylene Bottle Top Filter, Nylon Filter Material, 0.45m, 90mm, Non-sterile, 12/CS
SDP
Supplier:  CELLTREAT Scientific Products 229736
Encompass_Preferred
CELLTREAT Scientific Products 1000mL Polypropylene Bottle Top Filter with 0.45¿m Nylon Filter Material. Fits on standard GL 45 threaded glass vacuum bottles. Excellent chemical resistance offered with polypropylene housing as compared to polystyrene. High temperature range of 4?C to 37?C is suitable for many applications. Hydrophilic nylon membrane has excellent chemical compatibility with esters, bases and alcohols, and does not require pre-wetting. Large 90mm filter membrane diameter increases filtration speed and reduces clogging. Recommended operating pressure is 7.4psi. Maximum operating pressure is 9.8psi. Single piece Bottle Top Filter manufactured from virgin polypropylene. Membrane material - Nylon (polyamine - PA). Every case is printed with the lot number for quality traceability. Non-sterile. Individually packed in bags, 12/case.
Catalog No. 50-153-5088