Learn More
Ace Glass, Inc. 1000mL graduated pressure equalizing funnel, 24/40 top, PTFE metering plug
SDP
Supplier:  Ace Glass, Inc. 732014
Encompass_Preferred
1000ml graduated cylindrical pressure equalizing funnel with 24/40 top and bottom joints, drip stem, pressure equalizing arm, stopcock w/PTFE metering plug, and double scale graduations calibrated from stopcock.
Catalog No. 50-974-787