Learn More
Chemglass Life Sciences Flask, Round Bottom, 1000mL, Heavy Wall, 4-Neck, 29/42 CN - 24/40 SN's Angled 20°
SDP
Catalog No. CG153206
Encompass_Preferred
Flask, Round Bottom, 1000mL, Heavy Wall, 29/42 - 24/40, 4-Neck, Angled 20¿. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and 20¿ angled side necks.
Catalog No. CG153206 Supplier Chemglass Life Sciences Supplier No. CG153206
Flask, Round Bottom, 1000mL, Heavy Wall, 29/42 - 24/40, 4-Neck, Angled 20¿. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and 20¿ angled side necks.