Learn More
Chemglass Life Sciences 1000mL Reaction Vessel, Jacketed, 100mm Schott O-Ring Flange, GL-14 Inlet/Outlet, CG-911-A-10 Drain Valve
SDP
Supplier:  Chemglass Life Sciences CG193124
Encompass_Preferred
1000mL Reaction Vessel, Jacketed, 100mm Schott O-Ring Flange, GL-14 Inlet/Outlet, CG-911-A-10 Drain Valve. Similar to CG-1930, but with morton indentations to increase agitation during stirring. Supplied complete with two polypropylene hose connections and two screw caps. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum
Catalog No. CG193124